missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Aminobenzoyl)-L-glutamic acid diethyl ester, 99%, Thermo Scientific™

Product Code. 11475986
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475986 10 g 10g
11485986 50 g 50g
11495986 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475986 Supplier Thermo Scientific Alfa Aesar Supplier No. B22305.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13726-52-8
Molecular Formula C16H22N2O5
Molecular Weight (g/mol) 322.361
MDL Number MFCD00009617
InChI Key RJXFBLRRPYBPTM-ZDUSSCGKSA-N
Synonym n-4-aminobenzoyl-l-glutamic acid diethyl ester, diethyl n-4-aminobenzoyl-l-glutamate, diethyl n-p-aminobenzoyl-l-glutamate, 1,5-diethyl 2s-2-4-aminophenyl formamido pentanedioate, 4-abz-glu oet-oet, l-glutamic acid, n-4-aminobenzoyl-, diethyl ester, diethyl 4-aminobenzoyl-l-glutamate, diethyl p-aminobenzoyl-l-glutamate, s-diethyl 2-4-aminobenzamido pentanedioate, diethyl 2-4-aminobenzoyl amino pentanedioate #
PubChem CID 73002
IUPAC Name diethyl (2S)-2-[(4-aminobenzoyl)amino]pentanedioate
SMILES CCOC(=O)CCC(C(=O)OCC)NC(=O)C1=CC=C(C=C1)N

Specifications

Melting Point 139°C to 142°C
Quantity 10 g
Formula Weight 322.36
Optical Rotation 19° (c=1 Chloroform)
Percent Purity 99%
Chemical Name or Material N-(4-Aminobenzoyl)-L-glutamic acid diethyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.