missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Amino-2-chlorophenyl)-5-chloro-2-hydroxybenzamide, TRC

Product Code. 30474654
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30474654 50 mg
30476579 10 mg
30470258 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30474654 Supplier Toronto Research Chemicals Supplier No. TRCN28585850MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10558-45-9
Molecular Formula C13H10N2O2Cl2
Molecular Weight (g/mol) 297.14
IUPAC Name N-(4-amino-2-chlorophenyl)-5-chloro-2-hydroxybenzamide
SMILES Nc1ccc(NC(=O)c2cc(Cl)ccc2O)c(Cl)c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H10Cl2N2O2/c14-7-1-4-12(18)9(5-7)13(19)17-11-3-2-8(16)6-10(11)15/h1-6,18H,16H2,(H,17,19)
Formula Weight 296.0119
Chemical Name or Material N-(4-Amino-2-chlorophenyl)-5-chloro-2-hydroxybenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.