missing translation for 'onlineSavingsMsg'
Learn More

N-[4,6-Dichloro-5-(formylamino)-2-pyrimidinyl]acetamide, TRC

Product Code. 30382705
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382705 500 mg
30387177 250 mg
30387783 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382705 Supplier Toronto Research Chemicals Supplier No. TRCD434485500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136470-91-2
Molecular Formula C7 H6 Cl2 N4 O2
Molecular Weight (g/mol) 249.0541
Synonym Acetamide, N-[4,6-dichloro-5-(formylamino)-2-pyrimidinyl]-, N-(4,6-Dichloro-5-formamidopyrimidin-2-yl)acetamide, N-[4,6-Dichloro-5-(formylamino)-2-pyrimidinyl]acetamide
IUPAC Name N-(4,6-dichloro-5-formamidopyrimidin-2-yl)acetamide
SMILES CC(=O)Nc1nc(Cl)c(NC=O)c(Cl)n1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C7H6Cl2N4O2/c1-3(15)11-7-12-5(8)4(10-2-14)6(9)13-7/h2H,1H3,(H,10,14)(H,11,12,13,15)
Formula Weight 247.9868
Chemical Name or Material N-(4,6-Dichloro-5-formamidopyrimidin-2-yl)acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.