missing translation for 'onlineSavingsMsg'
Learn More

N-[4-[2-[(1-Methylethyl)amino]ethyl]phenyl]methanesulphonamide Hydrochloride, Mikromol™

Product Code. 18021570
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18021570 25 mg 25mg
18021270 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18021570 Supplier Mikromol Supplier No. MM0046.010025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-[4-[2-[(1-Methylethyl)amino]ethyl]phenyl]methanesulphonamide Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[4-[2-[(1-Methylethyl)amino]ethyl]phenyl]-methanesulphonamide Hydrochloride
CAS 16974-44-0
Analyte or Component Names N-[4-[2-[(1-Methylethyl)amino]ethyl]phenyl]-methanesulphonamide Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C12 H20 N2 O2 S . Cl H
Certifications/Compliance ISO 17025
Synonym Sotalol Hydrochloride Imp. A (EP), Sotalol Imp. A (EP), Sotalol USP Related Compound C, Sotalol USP RC C, N-[4-[2-[(1-Methylethyl)amino]ethyl]phenyl]-methanesulphonamide Hydrochloride, Sotalol Hydrochloride Imp. A (EP) as Hydrochloride, Sotalol Related Compound C as Hydrochloride, Sotalol Hydrochloride Impurity A as Hydrochloride, Sotalol Impurity A as Hydrochloride
InChI Formula InChI=1S/C12H20N2O2S.ClH/c1-10(2)13-9-8-11-4-6-12(7-5-11)14-17(3,15)16;/h4-7,10,13-14H,8-9H2,1-3H3;1H
SMILES Cl.CC(C)NCCc1ccc(NS(=O)(=O)C)cc1
IUPAC Name N-[4-[2-(propan-2-ylamino)ethyl]phenyl]methanesulfonamide;hydrochloride
Molecular Weight (g/mol) 292.83
Quantity 25 mg
Formula Weight 292.1012 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.