missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[4-[(1RS)-2-Hydroxy-1-[(1-methylethyl)amino]ethyl]phenyl]methanesulfonamide Hydrochloride, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
N-[4-[(1RS)-2-Hydroxy-1-[(1-methylethyl)amino]ethyl]phenyl]methanesulfonamide Hydrochloride
Specifications
Specifications
| Chemical Name or Material | N-[4-[(1RS)-2-Hydroxy-1-[(1-methylethyl)amino]ethyl]phenyl]methanesulfonamide Hydrochloride |
| CAS | 2724689-39-6 |
| Analyte or Component Names | N-[4-[(1RS)-2-Hydroxy-1-[(1-methylethyl)amino]ethyl]phenyl]methanesulfonamide Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H20 N2 O3 S . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Sotalol Hydrochloride Imp. D (EP), Sotalol Imp. D (EP), N-[4-[(1RS)-2-Hydroxy-1-[(1-methylethyl)amino]ethyl]phenyl]methanesulfonamide Hydrochloride, Sotalol Hydrochloride Impurity D as Hydrochloride, Sotalol Impurity D as Hydrochloride |
| InChI Formula | InChI=1S/C4H3N5O.H2O/c10-4-2-3(6-1-5-2)7-9-8-4;/h1H,(H2,5,6,7,8,10);1H4 |
| SMILES | Cl.CC(C)NC(CO)c1ccc(NS(=O)(=O)C)cc1 |
| IUPAC Name | N-[4-[2-hydroxy-1-(propan-2-ylamino)ethyl]phenyl]methanesulfonamide;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction