missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Hydroxypropyl) 4-hydroxy-2-nitroaniline, TRC

Product Code. 30399550
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399550 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399550 Supplier Toronto Research Chemicals Supplier No. TRCH9618655G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 92952-81-3
Molecular Formula C9H12N2O4
Quantity 5 g
Synonym 4-[(3-Hydroxypropyl)amino]-3-nitrophenol, HC Red BN, Jarocol Red BN, TM Red 2, Velsol Red BN
InChI Formula InChI=1S/C9H12N2O4/c12-5-1-4-10-8-3-2-7(13)6-9(8)11(14)15/h2-3,6,10,12-13H,1,4-5H2
SMILES OCCCNc1ccc(O)cc1[N+](=O)[O-]
IUPAC Name 4-(3-hydroxypropylamino)-3-nitrophenol
Molecular Weight (g/mol) 212.2
Formula Weight 212.0797
Chemical Name or Material N-(3-Hydroxypropyl) 4-hydroxy-2-nitroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.