missing translation for 'onlineSavingsMsg'
Learn More

N-[3-Hydroxy-1,1-bis(hydroxymethyl)-3-(4-octylphenyl)propyl]-acetamide Triethyamine (1:2), TRC

Product Code. 30401911
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401911 250 mg 250mg
30393236 50 mg 50mg
30393076 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401911 Supplier Toronto Research Chemicals Supplier No. TRCH714830250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[3-Hydroxy-1,1-bis(hydroxymethyl)-3-(4-octylphenyl)propyl]-acetamide Triethyamine (1:2)
Analyte or Component Names N-[3-Hydroxy-1,1-bis(hydroxymethyl)-3-(4-octylphenyl)propyl]-acetamide Triethyamine (1:2)
Shipping Condition Room Temperature
Molecular Formula C21H35NO4 . C 12H30N2
InChI Formula InChI=1S/C21H35NO4.2C6H15N/c1-3-4-5-6-7-8-9-18-10-12-19(13-11-18)20(26)14-21(15-23,16-24)22-17(2)25;2*1-4-7(5-2)6-3/h10-13,20,23-24,26H,3-9,14-16H2,1-2H3,(H,22,25);2*4-6H2,1-3H3
Solution Type Neat
SMILES OC(C1=CC=C(CCCCCCCC)C=C1)CC(CO)(NC(C)=O)CO.CCN(CC)CC.CCN(CC)CC
IUPAC Name N-(1,4-dihydroxy-2-(hydroxymethyl)-4-(4-octylphenyl)butan-2-yl)acetamide--triethylamine
Industry Type Pharmaceutical
Molecular Weight (g/mol) 567.89
Quantity 250 mg
Formula Weight 567.64
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.