missing translation for 'onlineSavingsMsg'
Learn More

n-(3-Formylphenyl)methanesulfonamide, ≥97%, Thermo Scientific™

Product Code. 10147252
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 55512-05-5
Molecular Formula C8H9NO3S
Molecular Weight (g/mol) 199.22
MDL Number MFCD03198179
InChI Key CBDSSTWZEANOCR-UHFFFAOYSA-N
Synonym n-3-formylphenyl methanesulfonamide, 3-methylsulfonamido benzaldehyde, methanesulfonamide, n-3-formylphenyl, 3-methylsulfonyl amino benzaldehyde, n-3-formylphenyl methanesulphonamide, m-methylsulfonylamino benzaldehyde, 3-methylsulfonylamino benzaldehyde, methanesulfonamide,n-3-formylphenyl, n-3-formyl-phenyl-methanesulfonamide
PubChem CID 2794787
IUPAC Name N-(3-formylphenyl)methanesulfonamide
SMILES CS(=O)(=O)NC1=CC=CC(C=O)=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 199.23
Percent Purity 97%
Chemical Name or Material N-(3-formylphenyl)methanesulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'