missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Formyl-2-pyridinyl)-2,2-dimethylpropanamide, ≥90%, Thermo Scientific™

Product Code. 10082124
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10082124 5 g 5g
10324292 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10082124 Supplier Thermo Scientific Maybridge Supplier No. MO00187DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 86847-64-5
Molecular Formula C11H14N2O2
Molecular Weight (g/mol) 206.245
MDL Number MFCD01830170
InChI Key ANABHCSYKASRRW-UHFFFAOYSA-N
Synonym n-3-formylpyridin-2-yl pivalamide, n-3-formyl-2-pyridinyl-2,2-dimethylpropanamide, n-3-formylpyridin-2-yl-2,2-dimethylpropanamide, n-3-formyl-pyridin-2-yl-2,2-dimethyl-propionamide, 2-2,2,2-trimethylacetamido pyridine-3-carboxaldehyde, 2-pivalylamino-pyridine-3-carboxyaldehyde, n-3-formyl-2-pyridinyl pivalamide, n-3-formyl 2-pyridyl-2,2-dimethylpropanamide, pubchem16526, acmc-20akc5
PubChem CID 736837
IUPAC Name N-(3-formylpyridin-2-yl)-2,2-dimethylpropanamide
SMILES CC(C)(C)C(=O)NC1=C(C=CC=N1)C=O

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Quantity 5 g
Formula Weight 206.24
Percent Purity 90%
Chemical Name or Material N-(3-Formyl-2-pyridinyl)-2, 2-dimethylpropanamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.