missing translation for 'onlineSavingsMsg'
Learn More

N-[3-Fluoro-4-[6-(2-methyl-2H-tetrazol-5-yl)-3-pyridinyl]phenyl]carbamic Acid Phenylmethyl Ester, TRC

Product Code. 30395215
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395215 25 mg 25mg
30408384 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30395215 Supplier Toronto Research Chemicals Supplier No. TRCF59363525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[3-Fluoro-4-[6-(2-methyl-2H-tetrazol-5-yl)-3-pyridinyl]phenyl]carbamic Acid Phenylmethyl Ester
CAS 1220910-89-3
Molecular Formula C21H17FN6O2
Quantity 25 mg
Synonym Tedizolid Impurity 68
InChI Formula InChI=1S/C21H17FN6O2/c1-28-26-20(25-27-28)19-10-7-15(12-23-19)17-9-8-16(11-18(17)22)24-21(29)30-13-14-5-3-2-4-6-14/h2-12H,13H2,1H3,(H,24,29)
SMILES FC1=C(C2=CC=C(C3=NN(C)N=N3)N=C2)C=CC(NC(OCC4=CC=CC=C4)=O)=C1
IUPAC Name benzyl (3-fluoro-4-(6-(2-methyl-2H-tetrazol-5-yl)pyridin-3-yl)phenyl)carbamate
Molecular Weight (g/mol) 404.4
Formula Weight 404.14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.