missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Bromopropyl)phthalimide, 98%

Product Code. 10142850 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10142850 100 g Glass bottle 100g
10162800 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10142850 Supplier Thermo Scientific Acros Supplier No. 107391000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5460-29-7 | C11H10BrNO2 | 268.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5460-29-7
Molecular Formula C11H10BrNO2
Molecular Weight (g/mol) 268.1
MDL Number MFCD00005904
InChI Key VKJCJJYNVIYVQR-UHFFFAOYSA-N
Synonym n-3-bromopropyl phthalimide, 2-3-bromopropyl isoindoline-1,3-dione, 1-phthalimido-3-bromopropane, 3-bromopropylphthalimide, n-3-bromopropylphthalimide, gamma-bromopropylphthalimide, 2-3-bromopropyl-1h-isoindole-1,3 2h-dione, 2-3-bromopropyl-2,3-dihydro-1h-isoindole-1,3-dione, phthalimide, n-3-bromopropyl, 1h-isoindole-1,3 2h-dione, 2-3-bromopropyl
PubChem CID 21611
IUPAC Name 2-(3-bromopropyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 70°C to 76°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 g
Beilstein 21, 462
Solubility Information Solubility in water: <1g/L. Other solubilities: soluble ca. 10g/L in organic solvents,soluble in ethanol and ether
Formula Weight 268.1
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material N-(3-Bromopropyl)phthalimide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • N-(3-Bromopropyl)phthalimide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.