missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Bromo-5-methyl-2-pyridyl)thiourea, 98+%, Thermo Scientific™

Catalog Number 15466417
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
866620-45-3
C7H8BrN3S
246.126
MFCD22682808
YISGTNGIIXFOQT-UHFFFAOYSA-N
n-3-bromo-5-methyl-2-pyridyl thiourea, 1-3-bromo-5-methylpyridin-2-yl thiourea
18506914
(3-bromo-5-methylpyridin-2-yl)thiourea
CC1=CC(=C(N=C1)NC(=S)N)Br
This item is not returnable. View return policy
866620-45-3
C7H8BrN3S
246.126
MFCD22682808
YISGTNGIIXFOQT-UHFFFAOYSA-N
n-3-bromo-5-methyl-2-pyridyl thiourea, 1-3-bromo-5-methylpyridin-2-yl thiourea
18506914
(3-bromo-5-methylpyridin-2-yl)thiourea
CC1=CC(=C(N=C1)NC(=S)N)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 210°C to 214°C
Quantity 250 mg
Formula Weight 246.12
Percent Purity ≥98%
Chemical Name or Material N-(3-Bromo-5-methyl-2-pyridyl)thiourea
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'