missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Aminopropyl)-N-methylcarbamic Acid tert-Butyl Ester Hydrochloride, TRC

Product Code. 30326600
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30326600 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326600 Supplier Toronto Research Chemicals Supplier No. TRCA6287105G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(3-Aminopropyl)-N-methylcarbamic Acid tert-Butyl Ester Hydrochloride
CAS 1188263-67-3
Molecular Formula C9 H18 N2 O2 . H Cl
Quantity 5 g
Synonym (3-Aminopropyl)methylcarbamic Acid 1,1-Dimethylethyl Ester Hydrochloride, 2-Methylpropan-2-yl N-(3-Aminopropyl)-N-methylcarbamate Hydrochloride, 3-(N-tert-Butoxycarbonyl-N-methylamino)propylamine Hydrochloride, tert-Butyl (3-Aminopropyl)(methyl)carbamate Hydrochloride, tert-Butyl N-(3-Aminopropyl)-N-(methyl)carbamate Hydrochloride
InChI Formula InChI=1S/C9H18N2O2.ClH/c1-9(2,3)13-8(12)11(4)7-5-6-10;/h6-7,10H2,1-4H3;1H
SMILES Cl.CN(C[C]CN)C(=O)OC(C)(C)C
IUPAC Name tert-butyl N-(3-aminopropyl)-N-methylcarbamate;hydrochloride
Molecular Weight (g/mol) 222.712
Formula Weight 222.114
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.