missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Aminopropyl)cyclohexylamine, 98%, Thermo Scientific™

Product Code. 11337646
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11337646 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11337646 Supplier Thermo Scientific Alfa Aesar Supplier No. L11250.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3312-60-5
Molecular Formula C9H20N2
Molecular Weight (g/mol) 156.273
MDL Number MFCD00003838
InChI Key ITZPOSYADVYECJ-UHFFFAOYSA-N
Synonym n-3-aminopropyl cyclohexylamine, n1-cyclohexylpropane-1,3-diamine, 1,3-propanediamine, n-cyclohexyl, n-cyclohexyl-1,3-propanediamine, cyclohexyl-1,3-propanediamine, 3-aminopropyl cyclohexylamine, n-cyclohexyl-1,3-diaminopropane, unii-f4xob09fkn, n-cyclohexyl-1,3-propylenediamine, n-cyclohexylpropylene 1,3-diamine
PubChem CID 18718
IUPAC Name N'-cyclohexylpropane-1,3-diamine
SMILES C1CCC(CC1)NCCCN

Specifications

Melting Point -18°C
Boiling Point 236°C
Flash Point 111°C (232°F)
Quantity 25 g
UN Number UN2735
Beilstein 741954
Formula Weight 156.27
Percent Purity 98%
Chemical Name or Material N-(3-Aminopropyl)cyclohexylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.