missing translation for 'onlineSavingsMsg'
Learn More

N-(3'-Aminopropyl)-2-pyrrolidinone, 95%

Product Code. 10219323 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Packaging:
Glass Bottle
Unit Size:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10219323 250 g Glass Bottle 250g
10426820 5 g Glass Bottle 5g
10436820 50 g Glass Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10219323 Supplier Thermo Scientific Acros Supplier No. 159602500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7663-77-6 | C7H14N2O | 142.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7663-77-6
Molecular Formula C7H14N2O
Molecular Weight (g/mol) 142.20
MDL Number MFCD00003201
InChI Key HJORCZCMNWLHMB-UHFFFAOYSA-N
Synonym 1-3-aminopropyl pyrrolidin-2-one, n-3'-aminopropyl-2-pyrrolidinone, 2-pyrrolidinone, 1-3-aminopropyl, 1-3-aminopropyl-2-pyrrolidone, n-3-aminopropyl-2-pyrrolidinone, 1-3-aminopropyl-2-pyrrolidinone, n-3-aminopropyl pyrrolidin-2-one, 1-3-amino-propyl-pyrrolidin-2-one, 1-3-aminoprop-1-yl pyrrolidin-2-one, zlchem 943
PubChem CID 82111
IUPAC Name 1-(3-aminopropyl)pyrrolidin-2-one
SMILES NCCCN1CCCC1=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Brown
Density 1.0100g/mL
Boiling Point 120.0°C to 123.0°C (1.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 1,5-Diazabicyclo(4.3.0.)non-5-ene: Balance (GC)
Packaging Glass Bottle
Refractive Index 1.4970 to 1.4990
Quantity 250 g
Specific Gravity 1.01
Formula Weight 142.2
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material N-(3'-Aminopropyl)-2-pyrrolidinone
Show More Show Less
compliance-icons
Product Identifier
  • N-(3'-Aminopropyl)-2-pyrrolidinone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.