missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(3-(Aminomethyl)benzyl)acetamidine Dihydrochloride, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | N-(3-(Aminomethyl)benzyl)acetamidine, Dihydrochloride |
| CAS | 214358-33-5 |
| Molecular Formula | C10 H15 N3 . 2 Cl H |
| Quantity | 5 mg |
| Synonym | 1400W dihydrochloride |
| InChI Formula | InChI=1S/C10H15N3.2ClH/c1-8(12)13-7-10-4-2-3-9(5-10)6-11;;/h2-5H,6-7,11H2,1H3,(H2,12,13);2*1H |
| SMILES | Cl.Cl.CC(=N)NCc1cccc(CN)c1 |
| IUPAC Name | N-[[3-(aminomethyl)phenyl]methyl]ethanimidamide;dihydrochloride |
| Molecular Weight (g/mol) | 250.17 |
| Formula Weight | 249.08 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction