missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Acetyl-4-hydroxyphenyl)butyramide, 97%

Product Code. 11482958
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482958 50 g 50g
11492958 250 g 250g
11472958 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482958 Supplier Thermo Scientific Alfa Aesar Supplier No. A18672.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 40188-45-2 | C12H15NO3 | 221.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40188-45-2
Molecular Formula C12H15NO3
Molecular Weight (g/mol) 221.26
MDL Number MFCD00798556
InChI Key FGWZEOPEZISTTR-UHFFFAOYSA-N
Synonym 2-acetyl-4-butyramidophenol, n-3-acetyl-4-hydroxyphenyl butanamide, n-3-acetyl-4-hydroxyphenyl butyramide, ac-hophe-oh, unii-14qjb65r4v, 3'-acetyl-4'-hydroxybutyranilide, butanamide, n-3-acetyl-4-hydroxyphenyl, 5'-butyramido-2'-hydroxyacetophenone, acebutolol impurity c, n-3-acetyl-4-hydroxyphenyl butanamid
PubChem CID 736331
IUPAC Name N-(3-acetyl-4-hydroxyphenyl)butanamide
SMILES CCCC(=O)NC1=CC=C(O)C(=C1)C(C)=O

Specifications

Melting Point 124°C to 126°C
Quantity 50 g
Formula Weight 221.26
Percent Purity 97%
Chemical Name or Material N-(3-Acetyl-4-hydroxyphenyl)butyramide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.