missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ N-(3,5-Dimethylphenyl)-3,5-difluorobenzenesulfonamide, 97%
Brand: Alfa Aesar™ H62082.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
N-(3,5-Dimethylphenyl)-3,5-difluorobenzenesulfonamide | |
297.32 | |
MFCD09394396 |
Specifications
N-(3,5-Dimethylphenyl)-3,5-difluorobenzenesulfonamide | |
97% | |
MFCD09394396 |
Solid | |
297.32 | |
1g |