missing translation for 'onlineSavingsMsg'
Learn More

N-(3,4-Dimethylphenyl)-4-fluorobenzenesulfonamide, 97%, Thermo Scientific™

Product Code. p-7036878
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492036 250 mg 250mg
15482036 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15492036 Supplier Thermo Scientific Alfa Aesar Supplier No. H55330.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-(3,4-Dimethylphenyl)-4-fluorobenzenesulfonamide is used in agrochemicals, reagents.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288154-65-4
Molecular Formula C14H14FNO2S
Molecular Weight (g/mol) 279.329
MDL Number MFCD00784407
InChI Key IJCSKFYHYFBPNB-UHFFFAOYSA-N
Synonym n-3,4-dimethylphenyl-4-fluorobenzenesulfonamide, cambridge id 5353558, 3,4-dimethylphenyl 4-fluorophenyl sulfonyl amine, n-3,4-dimethylphenyl-4-fluorobenzene-1-sulfonamide
PubChem CID 295161
IUPAC Name N-(3,4-dimethylphenyl)-4-fluorobenzenesulfonamide
SMILES CC1=C(C=C(C=C1)NS(=O)(=O)C2=CC=C(C=C2)F)C

Specifications

Quantity 250 mg
Formula Weight 279.33
Percent Purity 97%
Chemical Name or Material N-(3,4-Dimethylphenyl)-4-fluorobenzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.