missing translation for 'onlineSavingsMsg'
Learn More

N-[(3,4-Dimethoxy-2-pyridinyl)methyl] Pantoprazole(Mixture of 1 and 3 isomers), TRC

Product Code. 30373411
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30373411 100 mg
30385168 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30373411 Supplier Toronto Research Chemicals Supplier No. TRCD470960100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 957470-59-6
Molecular Formula C24H24F2N4O6S
Molecular Weight (g/mol) 534.53
Synonym 5-(Difluoromethoxy)-1-[(3,4-dimethoxy-2-pyridinyl)methyl]-2-[[(3,4-dimethoxy-2-pyridinyl)methyl]sulfinyl]-1H-benzimidazole, Pantoprazole Impurity
SMILES COc1ccnc(CS(=O)c2nc3cc(OC(F)F)ccc3n2Cc4nccc(OC)c4OC)c1OC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C24H24F2N4O6S/c1-32-19-7-9-27-16(21(19)34-3)12-30-18-6-5-14(36-23(25)26)11-15(18)29-24(30)37(31)13-17-22(35-4)20(33-2)8-10-28-17/h5-11,23H,12-13H2,1-4H3
Formula Weight 534.1385
Chemical Name or Material N-[(3,4-Dimethoxy-2-pyridinyl)methyl] Pantoprazole(Mixture of 1 and 3 isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.