missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(3,3-Dimethylbutyl)-L-a-aspartyl-L-phenylalanine Disodium Salt, TRC
Specifications
Specifications
| Molecular Formula | C19H26N2Na2O5 |
| Synonym | N-(3,3-Dimethylbutyl)-L-α-aspartyl-L-phenylalanine Disodium Salt, DMBAP Disodium Salt, USP Neotame Related Compound A Disodium Salt |
| SMILES | O=C(N[C@H](C(O[Na])=O)CC1=CC=CC=C1)[C@@H](NCCC(C)(C)C)CC(O[Na])=O |
| Molecular Weight (g/mol) | 408.4 |
| Quantity | 500 mg |
| Percent Purity | >95% |
| Shipping Condition | Room Temperature |
| Chemical Name or Material | N-(3,3-Dimethylbutyl)-L-alpha-aspartyl-L-phenylalanine Disodium Salt |
| Grade | HPLC |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?