missing translation for 'onlineSavingsMsg'
Learn More

N-[3-(2-Furyl)benzyl]-N-methylamine, ≥97%, Thermo Scientific™

Product Code. 10225364
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10225364 250 mg 250mg
10225844 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10225364 Supplier Thermo Scientific Maybridge Supplier No. CC46146CB
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 857284-27-6
Molecular Formula C12H13NO
Molecular Weight (g/mol) 187.242
MDL Number MFCD08435877
InChI Key WMCXJYGZLCREMM-UHFFFAOYSA-N
Synonym n-3-2-furyl benzyl-n-methylamine, 3-furan-2-yl phenyl methyl methyl amine, 1-3-furan-2-yl phenyl-n-methylmethanamine, 3-2-furyl phenyl methyl methylamine, benzenemethanamine,3-2-furanyl-n-methyl
PubChem CID 18525790
IUPAC Name 1-[3-(furan-2-yl)phenyl]-N-methylmethanamine
SMILES CNCC1=CC=CC(=C1)C2=CC=CO2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 187.24
Percent Purity 97%
Chemical Name or Material N-[3-(2-furyl)benzyl]-N-methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.