missing translation for 'onlineSavingsMsg'
Learn More

N-[(2Z)-Piperazin-2-ylidene]-2,2,2-trifluoroacetohydrazide, TRC

Product Code. 30421027
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30421027 1 g 1g
30412857 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30421027 Supplier Toronto Research Chemicals Supplier No. TRCP4810001G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 849832-73-1
Molecular Formula C6 H9 F3 N4 O
Molecular Weight (g/mol) 210.16
Synonym Acetic acid, 2,2,2-trifluoro-, (2Z)-2-(2-piperazinylidene)hydrazide, Acetic acid, trifluoro-, (2Z)-2-(2-piperazinylidene)hydrazide (9CI), Acetic acid, trifluoro-, (2Z)-piperazinylidenehydrazide, N-[(2Z)-Piperazin-2-ylidene]-2,2,2-trifluoroacetohydrazide, N'-[(2Z)-Piperazin-2-ylidene]trifluoroacetohydrazide
IUPAC Name 2,2,2-trifluoro-N-[(Z)-piperazin-2-ylideneamino]acetamide
SMILES FC(F)(F)C(=O)N\N=C/1\CNCCN1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H9F3N4O/c7-6(8,9)5(14)13-12-4-3-10-1-2-11-4/h10H,1-3H2,(H,11,12)(H,13,14)
Formula Weight 210.0728
Chemical Name or Material N-[(2Z)-Piperazin-2-ylidene]-2,2,2-trifluoroacetohydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.