missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-[(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyryl]-L-leucine, 97%

Product Code. 10430432 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10430432 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10430432 Supplier Thermo Scientific Chemicals Supplier No. 333915000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 58970-76-6 | C16H24N2O4 | 308.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 58970-76-6
CAS Min % 96.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C16H24N2O4
MDL Number MFCD00083262
Quantity 500 mg
Merck Index 15, 10024
Synonym bestatin, ubenimex, ubenimexum latin, ubenimex inn:jan, bestatin;ubenimex, s-2-2s,3r-3-amino-2-hydroxy-4-phenylbutanamido-4-methylpentanoic acid, ubenimex bestatin, chembl29292, 2s,3r-3-amino-2-hydroxy-4-phenylbutanoyl-l-leucine, 3-r-amino-2-s-hydroxy-4-phenylbutanoyl-s-leucine
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble methanol, ethanol,, soluble in acetic acid, aq. hcl, alkaline solution
InChI Key VGGGPCQERPFHOB-MCIONIFRSA-N
SMILES CC(C)C[C@@H](NC(=O)[C@@H](O)[C@H](N)CC1=CC=CC=C1)C(O)=O
IUPAC Name (2S)-2-[[(2S,3R)-3-amino-2-hydroxy-4-phenylbutanoyl]amino]-4-methylpentanoic acid
Molecular Weight (g/mol) 308.38
PubChem CID 72172
Formula Weight 308.38
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-[(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyryl]-L-leucine, 97%
Show More Show Less
compliance-icons
Product Identifier
  • N-¢(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyryl!-L-leucine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.