missing translation for 'onlineSavingsMsg'
Learn More

N-((2S,3R)-1-(((R)-1-(1,3,6,2-Dioxazaborocan-2-yl)-3-methylbutyl)amino)-3-hydroxy-1-oxobutan-2-yl)-6-phenylpicolinamide, TRC

Product Code. 30374427
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30374427 50 mg 50mg
30381873 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30374427 Supplier Toronto Research Chemicals Supplier No. TRCD48595050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-((2S,3R)-1-(((R)-1-(1,3,6,2-Dioxazaborocan-2-yl)-3-methylbutyl)amino)-3-hydroxy-1-oxobutan-2-yl)-6-phenylpicolinamide
Analyte or Component Names N-((2S,3R)-1-(((R)-1-(1,3,6,2-Dioxazaborocan-2-yl)-3-methylbutyl)amino)-3-hydroxy-1-oxobutan-2-yl)-6-phenylpicolinamide
Shipping Condition Room Temperature
Molecular Formula C25H35BN4O5
InChI Formula InChI=1S/C25H35BN4O5/c1-17(2)16-22(26-34-14-12-27-13-15-35-26)29-25(33)23(18(3)31)30-24(32)21-11-7-10-20(28-21)19-8-5-4-6-9-19/h4-11,17-18,22-23,27,31H,12-16H2,1-3H3,(H,29,33)(H,30,32)/t18-,22-,23+/m1/s1
Solution Type Neat
SMILES O=C([C@H]([C@@H](C)O)NC(C1=CC=CC(C2=CC=CC=C2)=N1)=O)N[C@@H](CC(C)C)B3OCCNCCO3
IUPAC Name N-((2S,3R)-1-(((R)-1-(1,3,6,2-dioxazaborocan-2-yl)-3-methylbutyl)amino)-3-hydroxy-1-oxobutan-2-yl)-6-phenylpicolinamide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 482.38
Quantity 50 mg
Formula Weight 482.27
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.