missing translation for 'onlineSavingsMsg'
Learn More

N-(23-Amino-3,6,9,12,15,18,21-heptaoxatricos-1-yl)-2-[2,6-bis(1,1-dimethylethyl)-4-[6-(4-methyl-1-piperazinyl)[2,6′-bi-1H-benzimidazol]-2′-yl]phenoxy]acetamide, TRC

Product Code. 30314910
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30314910 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314910 Supplier Toronto Research Chemicals Supplier No. TRCA14154050MG
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(23-Amino-3,6,9,12,15,18,21-heptaoxatricos-1-yl)-2-[2,6-bis(1,1-dimethylethyl)-4-[6-(4-methyl-1-piperazinyl)[2,6'-bi-1H-benzimidazol]-2'-yl]phenoxy]acetamide
CAS 1071790-45-8
Molecular Formula C51H76N8O9
Quantity 50 mg
InChI Formula InChI=1S/C51H76N8O9/c1-50(2,3)40-32-38(49-55-42-10-8-37(34-44(42)56-49)48-54-43-11-9-39(35-45(43)57-48)59-16-14-58(7)15-17-59)33-41(51(4,5)6)47(40)68-36-46(60)53-13-19-62-21-23-64-25-27-66-29-31-67-30-28-65-26-24-63-22-20-61-18-12-52/h8-11,32-35H,12-31,36,52H2,1-7H3,(H,53,60)(H,54,57)(H,55,56)
SMILES CN1CCN(C2=CC=C3N=C(C4=CC=C5C(N=C(C6=CC(C(C)(C)C)=C(OCC(NCCOCCOCCOCCOCCOCCOCCOCCN)=O)C(C(C)(C)C)=C6)N5)=C4)NC3=C2)CC1
IUPAC Name N-(23-amino-3,6,9,12,15,18,21-heptaoxatricosyl)-2-(2,6-di-tert-butyl-4-(6-(4-methylpiperazin-1-yl)-1H,1'H-[2,5'-bibenzo[d]imidazol]-2'-yl)phenoxy)acetamide
Molecular Weight (g/mol) 945.22
Formula Weight 944.57
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.