missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Pyridyl)bis(trifluoromethanesulfonimide), 97%

Product Code. 10350534 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10350534 5 g Glass bottle 5g
10742774 1 g Glass bottle 1g
10646704 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10350534 Supplier Thermo Scientific Acros Supplier No. 387650050
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
This item is not returnable. View return policy

CAS: 145100-50-1 | C7H4F6N2O4S2 | 358.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145100-50-1
Molecular Formula C7H4F6N2O4S2
Molecular Weight (g/mol) 358.23
MDL Number MFCD00191834
InChI Key DXLQEJHUQKKSRB-UHFFFAOYSA-N
Synonym 2-n,n-bis trifluoromethylsulfonyl amino pyridine, 1,1,1-trifluoro-n-pyridin-2-yl-n-trifluoromethyl sulfonyl methanesulfonamide, n-pyridin-2-yl triflimide, 2-n,n-bis trifluoromethyl sulphonylamino pyridine, n-2-pyridyl triflimide, n-2-pyridyl bis trifluoromethanesulfonimide, pyridin-2-amine, n,n-bis trifluoromethylsulfonyl, 2-bis trifluoromethylsulfonyl amino pyridine, 1,1,1-trifluoro-n-pyridin-2-yl-n-trifluoromethanesulfonylmethanesulfonamide, n,n-bis trifluoromethylsulfonyl-2-pyridylamine
PubChem CID 534172
IUPAC Name 1,1,1-trifluoro-N-pyridin-2-yl-N-(trifluoromethylsulfonyl)methanesulfonamide
SMILES FC(F)(F)S(=O)(=O)N(C1=CC=CC=N1)S(=O)(=O)C(F)(F)F

Specifications

Boiling Point 97.0°C (2.0 mmHg)
Melting Point 40.0°C to 43.0°C
Color White
CAS Min % 96.0
CAS Max % 100.0
Physical Form Crystalline Powder and/or Crystals
Quantity 5 g
Assay Percent Range 96% min. (HPLC)
Fieser 17,293
Flash Point 110°C
Formula Weight 358.24
Percent Purity 97%
Infrared Spectrum Authentic
Packaging Glass bottle
Density 110g/mL
Chemical Name or Material N-(2-Pyridyl)bis(trifluoromethanesulfonimide)
Show More Show Less
compliance-icons
Product Identifier
  • N-(2-Pyridyl)bis(trifluoroethanesulfonimide)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.