missing translation for 'onlineSavingsMsg'
Learn More

N-[2-[(Methylamino)carbonyl]benzoyl] Amlodipine, TRC

Product Code. 17982065
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17982065 50 mg 50mg
17934325 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17982065 Supplier Toronto Research Chemicals Supplier No. TRCM28580050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 721958-72-1
Molecular Formula C29 H32 Cl N3 O7
Molecular Weight (g/mol) 570.03
Synonym 3-Ethyl 5-Methyl (4RS)-4-(2-Chlorophenyl)-6-methyl-2-[[2-[[2-(methylcarbamoyl)benzoyl]amino]ethoxy]methyl]-1,4-dihydropyridine-3,5-dicarboxylate, Amlodipine Besilate Imp. B (EP)
IUPAC Name 5-O-ethyl 3-O-methyl 4-(2-chlorophenyl)-2-methyl-6-[2-[[2-(methylcarbamoyl)benzoyl]amino]ethoxymethyl]-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(COCCNC(=O)c2ccccc2C(=O)NC)NC(=C(C1c3ccccc3Cl)C(=O)OC)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C29H32ClN3O7/c1-5-40-29(37)25-22(16-39-15-14-32-27(35)19-11-7-6-10-18(19)26(34)31-3)33-17(2)23(28(36)38-4)24(25)20-12-8-9-13-21(20)30/h6-13,24,33H,5,14-16H2,1-4H3,(H,31,34)(H,32,35)
Formula Weight 569.1929
Percent Purity >95%
Chemical Name or Material 3-ethyl 5-methyl (4RS)-4-(2-chlorophenyl)-6-methyl-2-[(2-[(2-(methylcarbamoyl)benzoyl)amino]ethoxy)methyl]-1,4-dihydropyridine-3,5-dicarboxylate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.