missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Methoxyphenyl)thiourea, 94%, Thermo Scientific™

Codice prodotto. 15488939
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
missing translation for 'unitSize'
5g
Questo articolo non è restituibile. Consulta la politica di reso

Codice prodotto. 15488939

missing translation for 'mfr': Thermo Scientific Alfa Aesar L11840.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

Questo articolo non è restituibile. Consulta la politica di reso

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifica

CAS 1516-37-6
Melting Point 152°C to 156°C
Odor Odorless
Molecular Formula C8H10N2OS
MDL Number MFCD00022162
Quantity 5 g
UN Number UN2811
Beilstein 778949
Synonym 2-methoxyphenyl thiourea, 1-2-methoxyphenyl thiourea, o-methoxyphenylthiourea, n-2-methoxyphenyl thiourea, n-o-methoxyphenyl thiourea, 1-2-methoxyphenyl-2-thiourea, 1-o-methoxyphenyl thiourea, urea, 1-o-methoxyphenyl-2-thio, 1-o-methoxyphenyl-2-thiourea
InChI Key HXCHZMHFZXNFIX-UHFFFAOYSA-N
SMILES COC1=CC=CC=C1NC(=S)N
IUPAC Name (2-methoxyphenyl)thiourea
Molecular Weight (g/mol) 182.241
PubChem CID 722774
Formula Weight 182.25
Percent Purity 94%
Chemical Name or Material N-(2-Methoxyphenyl)thiourea
Vedi altri risultati Mostra meno risultati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.