missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Indol-3-ylethyl)-succinamic Acid Methyl Ester, TRC

Product Code. 30398757
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398757 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398757 Supplier Toronto Research Chemicals Supplier No. TRCI5765055MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(2-Indol-3-ylethyl)-succinamic Acid Methyl Ester
CAS 92870-51-4
Molecular Formula C8 H12 N3 O5 P
Quantity 5 mg
Synonym NSC 149841
InChI Formula InChI=1S/C8H12N3O5P/c12-7(13)6-3-11-5(4-17(14,15)16)1-2-9-8(11)10-6/h3,5H,1-2,4H2,(H,9,10)(H,12,13)(H2,14,15,16)
SMILES OC(=O)C1=CN2C(CP(=O)(O)O)CCN=C2N1
IUPAC Name 5-(phosphonomethyl)-1,5,6,7-tetrahydroimidazo[1,2-a]pyrimidine-2-carboxylic acid
Molecular Weight (g/mol) 261.172
Formula Weight 261.051
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.