missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)pyrrolidine, 95%

Product Code. 10132680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10132680 100 g Glass bottle 100g
10336850 25 g Glass bottle 25g
10391044 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10132680 Supplier Thermo Scientific Acros Supplier No. 121201000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2955-88-6 | C6H13NO | 115.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2955-88-6
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.18
MDL Number MFCD00003181
InChI Key XBRDBODLCHKXHI-UHFFFAOYSA-N
Synonym 1-pyrrolidineethanol, n-2-hydroxyethyl pyrrolidine, 1-2-hydroxyethyl pyrrolidine, 2-pyrrolidin-1-yl ethanol, epolamine, 2-pyrrolidinoethanol, pyrrolidinoethanol, hydroxyethylpyrrolidine, 2-1-pyrrolidinyl ethanol, 2-pyrrolidin-1-yl ethan-1-ol
PubChem CID 76288
ChEBI CHEBI:48293
IUPAC Name 2-pyrrolidin-1-ylethanol
SMILES OCCN1CCCC1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Brown to Yellow
Density 0.9800g/mL
Boiling Point 79°C to 81°C (13.0 mmHg)
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4713 to 1.4743
Quantity 100 g
Beilstein 20, II, 5
Specific Gravity 0.98
Formula Weight 115.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material N-(2-Hydroxyethyl)pyrrolidine
Show More Show Less
compliance-icons
Product Identifier
  • N-(2-Hydroxyethyl)pyrrolidine
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.