missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)phthalimide, 98+%

Product Code. 11492397
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11492397 500 g 500g
11482397 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11492397 Supplier Thermo Scientific Alfa Aesar Supplier No. A14259.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3891-07-4 | C17H22NO6 | 336.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3891-07-4
MDL Number MFCD00005903

Specifications

Melting Point 126°C to 129°C
Molecular Formula C17H22NO6
Quantity 500 g
Beilstein 147214
Synonym z-glu otbu-oh, cbz-glu obut-oh, cbz-l-glu otbu-oh, s-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, n-cbz-l-glutamic acid 5-tert-butyl ester, n-benzyloxycarbonyl-l-glutamic acid 5-tert-butyl ester, 2s-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, 2s-4-tert-butyl oxycarbonyl-2-phenylmethoxy carbonylamino butanoic acid, 5-tert-butyl n-carbobenzoxy-l-glutamate, pubchem6261
InChI Key GLMODRZPPBZPPB-ZDUSSCGKSA-M
SMILES CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
IUPAC Name (2S)-5-[(2-methylpropan-2-yl)oxy]-5-oxo-2-(phenylmethoxycarbonylamino)pentanoic acid
Molecular Weight (g/mol) 336.37
PubChem CID 107472
Formula Weight 191.19
Percent Purity ≥98%
Chemical Name or Material N-(2-Hydroxyethyl)phthalimide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.