missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)phthalimide, 98+%

Product Code. 11482397
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11482397 100 g 100g
11492397 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11482397 Supplier Thermo Scientific Alfa Aesar Supplier No. A14259.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3891-07-4 | C17H22NO6 | 336.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3891-07-4
MDL Number MFCD00005903

Specifications

Melting Point 126°C to 129°C
Molecular Formula C14H19NO3
Quantity 100 g
Beilstein 147214
Synonym 2-1,2,2-trimethylpropylcarbamoyl benzoic acid, 2-3,3-dimethylbutan-2-yl carbamoyl benzoic acid, 2-1,2,2-trimethylpropyl amino carbonyl benzoic acid, n-pinacolylphthalamic acid, 2-3,3-dimethylbutan-2-ylcarbamoyl benzoic acid, +-n-pinacolylphthalamic acid
InChI Key QUZGNIZLRAGRHB-UHFFFAOYNA-N
SMILES CC(C(C)(C)C)NC(=O)C1=CC=CC=C1C(=O)O
IUPAC Name 2-(3,3-dimethylbutan-2-ylcarbamoyl)benzoic acid
Molecular Weight (g/mol) 249.31
PubChem CID 3354762
Formula Weight 191.19
Percent Purity ≥98%
Chemical Name or Material N-(2-Hydroxyethyl)phthalimide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.