missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)-P,P-bisaziridinyl Thiophosphamide, TRC

Product Code. 30394488
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394488 25 mg
30391375 5 mg
30405962 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394488 Supplier Toronto Research Chemicals Supplier No. TRCH94249525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121258-29-5
Molecular Formula C6H14N3OPS
Molecular Weight (g/mol) 207.23
Synonym P,P-Bis(1-aziridinyl)-N-(2-hydroxyethyl)phosphinothioic Amide
IUPAC Name 2-[bis(aziridin-1-yl)phosphinothioylamino]ethanol
SMILES S=P(N1CC1)(N2CC2)NCCO

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C6H14N3OPS/c10-6-1-7-11(12,8-2-3-8)9-4-5-9/h10H,1-6H2,(H,7,12)
Formula Weight 207.0595
Chemical Name or Material N-(2-Hydroxyethyl)-P,P-bisaziridinyl Thiophosphamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.