missing translation for 'onlineSavingsMsg'
Learn More

N-2-(Hydroxyethyl)-L-valine-d4 (Technical grade), TRC

Product Code. 30391476
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391476 5 mg 5mg
30408288 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30391476 Supplier Toronto Research Chemicals Supplier No. TRCH9423025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120398-50-7
Molecular Formula C7H11D4NO3
Molecular Weight (g/mol) 165.22
Synonym N-2-(Hydroxyethyl)-L-Valine-D4
IUPAC Name (2S)-3-methyl-2-[(1,1,2,2-tetradeuterio-2-hydroxyethyl)amino]butanoic acid
SMILES [2H]C([2H])(O)C([2H])([2H])N[C@@H](C(C)C)C(=O)O

Specifications

InChI Formula InChI=1S/C7H15NO3/c1-5(2)6(7(10)11)8-3-4-9/h5-6,8-9H,3-4H2,1-2H3,(H,10,11)/t6-/m0/s1/i3D2,4D2
Quantity 5 mg
Formula Weight 165.1303
Shipping Condition Room Temperature
Chemical Name or Material 2-Hydroxyethyl-1,1,2,2-d4 Valine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.