missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)ethylenediamine, 99%

Product Code. 11408790
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408790 100 g 100g
11429792 500 g 500g
11439792 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11408790 Supplier Thermo Scientific Alfa Aesar Supplier No. A10833.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-41-1 | C4H12N2O | 104.15 g/mol

It is used as an important intermediate in the manufacture of lube oil additives, fuel additives, chelating agents, surfactants and fabric softeners among other applications. N-(2-Hydroxyethyl)ethylenediamine [N-(2-Aminoethyl)ethanolamine] is used to study the aerobic biodecomposition of amines in hypersaline wastewaters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an important intermediate in the manufacture of lube oil additives, fuel additives, chelating agents, surfactants and fabric softeners among other applications. N-(2-Hydroxyethyl)ethylenediamine [N-(2-Aminoethyl)ethanolamine] is used to study the aerobic biodecomposition of amines in hypersaline wastewaters.

Solubility
Fully miscible in water.

Notes
Air Sensitive & Hygroscopic. Store under inert gas. Keep away from incompatibles.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-41-1
Molecular Formula C4H12N2O
Molecular Weight (g/mol) 104.15
MDL Number MFCD00008170
InChI Key LHIJANUOQQMGNT-UHFFFAOYSA-N
Synonym 2-2-aminoethylamino ethanol, n-2-hydroxyethyl ethylenediamine, aminoethylethanolamine, n-2-aminoethyl ethanolamine, ethanol, 2-2-aminoethyl amino, 2-2-aminoethyl amino ethanol, monoethanolethylenediamine, n-hydroxyethyl ethylenediamine, n-aminoethyl ethanolamine, 2-aminoethyl ethanolamine
PubChem CID 8112
IUPAC Name 2-(2-aminoethylamino)ethanol
SMILES NCCNCCO

Specifications

Melting Point -28°C
Density 1.029
pH 11.8
Boiling Point 238°C to 240°C
Flash Point 125°C (257°F)
Odor Amine-like
Refractive Index 1.4861
Quantity 100 g
UN Number UN2735
Beilstein 506012
Sensitivity Air sensitive; Hygroscopic
Solubility Information Fully miscible in water.
Formula Weight 104.15
Percent Purity 99%
Chemical Name or Material N- (2-Hydroxyethyl)ethylenediamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N-(2-Hydroxyethyl)ethylenediamine
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: N-(2-Hydroxyethyl)ethylenediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.