missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)adenosine, TRC

Product Code. 30396235
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396235 50 mg -
30408632 25 mg 25mg
30408633 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396235 Supplier Toronto Research Chemicals Supplier No. TRCH99812050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4338-48-1
Molecular Formula C12H17N5O5
Molecular Weight (g/mol) 311.29
Synonym 6-Hydroxyethyladenosine, 6-[(2-Hydroxyethyl)amino]-9-beta-D-ribofuranosylpurine, N6-(2-Hydroxyethyl)adenosine, NSC 54251
IUPAC Name (2R,3R,4S,5R)-2-[6-(2-hydroxyethylamino)purin-9-yl]-5-(hydroxymethyl)oxolane-3,4-diol
SMILES OCCNc1ncnc2c1ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H17N5O5/c18-2-1-13-10-7-11(15-4-14-10)17(5-16-7)12-9(21)8(20)6(3-19)22-12/h4-6,8-9,12,18-21H,1-3H2,(H,13,14,15)/t6-,8-,9-,12-/m1/s1
Formula Weight 311.123
Chemical Name or Material N-(2-Hydroxyethyl)adenosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.