missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Guanidinoethyl)-5-isoquinolinesulfonamide Hydrochloride, TRC

Product Code. 30402733
Change view
Click to view available options
Quantity:
5 mg
25 mg
250 mg
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402733 250 mg 250mg
30394261 25 mg -
30389139 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402733 Supplier Toronto Research Chemicals Supplier No. TRCG835000250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92564-34-6
Molecular Formula C12 H15 N5 O2 S . Cl H
Molecular Weight (g/mol) 329.81
IUPAC Name 1-[2-(isoquinolin-5-ylsulfonylamino)ethyl]guanidine;hydrochloride
SMILES Cl.NC(=N)NCCNS(=O)(=O)c1cccc2cnccc12

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H15N5O2S.ClH/c13-12(14)16-6-7-17-20(18,19)11-3-1-2-9-8-15-5-4-10(9)11;/h1-5,8,17H,6-7H2,(H4,13,14,16);1H
Formula Weight 329.0713
Chemical Name or Material N-(2-Guanidinoethyl)-5-isoquinolinesulfonamide, Hydrochlorid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.