missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Ethylhexyl)-4-nitrobenzamide, 97%, Thermo Scientific™

Product Code. 15437328
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154702-08-6
Molecular Formula C15H22N2O3
Molecular Weight (g/mol) 278.352
MDL Number MFCD00709791
InChI Key XYLYSNQGXATQBX-UHFFFAOYSA-N
Synonym n-2-ethylhexyl-4-nitrobenzamide
PubChem CID 4139944
IUPAC Name N-(2-ethylhexyl)-4-nitrobenzamide
SMILES CCCCC(CC)CNC(=O)C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Quantity 250 mg
Formula Weight 278.35
Percent Purity 97%
Chemical Name or Material N-(2-Ethylhexyl)-4-nitrobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'