missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(2-Chloropyridin-4-yl)methyl]-N-methylamine, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 5 g |
Synonym | n-2-chloropyridin-4-yl methyl-n-methylamine, 2-chloropyridin-4-yl methyl methyl amine, 1-2-chloropyridin-4-yl-n-methylmethanamine, 2-chloro-4-methylamino methyl pyridine, 4-pyridinemethanamine,2-chloro-n-methyl, 2-chloro 4-pyridyl methyl methylamine, 4-pyridinemethanamine, 2-chloro-n-methyl, 2-chloro-n-methyl-4-pyridinemethanamine, 2-chloro-pyridin-4-ylmethyl-methyl-amine |
Formula Weight | 156.62 |
Percent Purity | 97% |
Chemical Name or Material | N-[(2-Chloropyridin-4-yl)methyl]-N-methylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'