missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Chloroethyl)pyrrolidine hydrochloride, 98%

Product Code. 10134312 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10134312 25 g Glass bottle 25g
10651745 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10134312 Supplier Thermo Scientific Acros Supplier No. 109220250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7250-67-1 | C6H12ClN·HCl | 170.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7250-67-1
Molecular Formula C6H12ClN·HCl
Molecular Weight (g/mol) 170.08
MDL Number MFCD00012718
InChI Key FSNGFFWICFYWQC-UHFFFAOYSA-N
Synonym 1-2-chloroethyl pyrrolidine hydrochloride, n-2-chloroethyl pyrrolidine hydrochloride, 2-chloroethylpyrrolidine hydrochloride, 2-pyrrolidinoethyl chloride hydrochloride, pyrrolidine, 1-2-chloroethyl-, hydrochloride, unii-yyq1h343r5, pyrrolidinoethyl chloride, hydrochloride, 1-2-chloroethyl pyrrolidine.hcl, 1-2-chloroethyl pyrrolidine hcl, 2-chloroethylpyrrolidinehydrochloride
PubChem CID 81668
IUPAC Name 1-(2-chloroethyl)pyrrolidine;hydrochloride
SMILES C1CCN(C1)CCCl.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 171.0°C to 175.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 20, II, 4
Solubility Information Solubility in water: soluble
Formula Weight 170.08
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material N-(2-Chloroethyl)pyrrolidine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • N-(2-Chloroethyl)pyrrolidine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.