Learn More
N-(2-Chloroethyl)morpholine hydrochloride, 99%, Thermo Scientific Chemicals
£13.40 - £129.00
Chemical Identifiers
CAS | 3647-69-6 |
---|---|
Molecular Formula | C6H12ClNO·HCl |
Molecular Weight (g/mol) | 186.08 |
MDL Number | MFCD00012797 |
InChI Key | NBJHDLKSWUDGJG-UHFFFAOYSA-N |
Synonym | 4-2-chloroethyl morpholine hydrochloride, n-2-chloroethyl morpholine hydrochloride, 2-morpholinoethylchloride hydrochloride, morpholine, 4-2-chloroethyl-, hydrochloride, 4-2-chloroethyl morpholine hcl, chloromorpholinoethane hydrochloride, 2-chloroethyl morpholine hydrochloride, 2-morpholinoethyl chloride hydrochloride, 4-2-chloroethyl-morpholine hydrochloride, 4-2-chloroethyl morpholinium chloride |
PubChem CID | 77210 |
IUPAC Name | 4-(2-chloroethyl)morpholine;hydrochloride |
SMILES | C1COCCN1CCCl.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10616872
|
Thermo Scientific Acros
109200050 |
5 g |
£13.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10427100
|
Thermo Scientific Acros
109201000 |
100 g |
£31.20
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10050850
|
Thermo Scientific Acros
109205000 |
500 g |
£129.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3647-69-6 | |
186.08 | |
NBJHDLKSWUDGJG-UHFFFAOYSA-N | |
77210 | |
C1COCCN1CCCl.Cl |
C6H12ClNO·HCl | |
MFCD00012797 | |
4-2-chloroethyl morpholine hydrochloride, n-2-chloroethyl morpholine hydrochloride, 2-morpholinoethylchloride hydrochloride, morpholine, 4-2-chloroethyl-, hydrochloride, 4-2-chloroethyl morpholine hcl, chloromorpholinoethane hydrochloride, 2-chloroethyl morpholine hydrochloride, 2-morpholinoethyl chloride hydrochloride, 4-2-chloroethyl-morpholine hydrochloride, 4-2-chloroethyl morpholinium chloride | |
4-(2-chloroethyl)morpholine;hydrochloride |
Specifications
3647-69-6 | |
100.0 | |
White to Beige | |
Authentic | |
Glass bottle | |
MFCD00012797 | |
Solubility in water: soluble | |
C1COCCN1CCCl.Cl | |
186.08 | |
186.08 | |
Hygroscopic Crystalline Powder |
98.5 | |
182°C to 185°C | |
1% max. (80°C) | |
99% | |
C6H12ClNO·HCl | |
4-2-chloroethyl morpholine hydrochloride, n-2-chloroethyl morpholine hydrochloride, 2-morpholinoethylchloride hydrochloride, morpholine, 4-2-chloroethyl-, hydrochloride, 4-2-chloroethyl morpholine hcl, chloromorpholinoethane hydrochloride, 2-chloroethyl morpholine hydrochloride, 2-morpholinoethyl chloride hydrochloride, 4-2-chloroethyl-morpholine hydrochloride, 4-2-chloroethyl morpholinium chloride | |
NBJHDLKSWUDGJG-UHFFFAOYSA-N | |
4-(2-chloroethyl)morpholine;hydrochloride | |
77210 | |
99% | |
N-(2-Chloroethyl)morpholine hydrochloride, 99% |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.
Harmful in contact with skin.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 222-881-2
missing translation for 'rtecsNumber' : QE0260000
missing translation for 'tsca' : TSCA
RUO – Research Use Only