missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Chloroethyl)acetamide, 98%

Product Code. 10062240 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10062240 50 g 50g
10573931 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10062240 Supplier Thermo Scientific Acros Supplier No. 184520500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7355-58-0 | C4H8ClNO | 121.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7355-58-0
Molecular Formula C4H8ClNO
Molecular Weight (g/mol) 121.57
MDL Number MFCD00000965
InChI Key HSKNJSHFPPHTAQ-UHFFFAOYSA-N
Synonym n-2-chloroethyl acetamide, acetamide, n-2-chloroethyl, n-2-chloro-ethyl-acetamide, n-acetyl-2-chloroethylamine, n 2-chloroethyl acetamide, n-acetyl-ss-chlorathylamin, acmc-1bi05, n-2-chloroethyl-acetamide, 4-04-00-00449 beilstein handbook reference
PubChem CID 81815
IUPAC Name N-(2-chloroethyl)acetamide
SMILES CC(=O)NCCCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2000g/mL
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CONHCH2CH2Cl
Refractive Index 1.4770 to 1.48
Quantity 50 g
Specific Gravity 1.2
Formula Weight 121.57
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material N-(2-Chloroethyl)acetamide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • N-(2-Chloroethyl)acetamide
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.