missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(2-Chloroethyl)acetamide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Color | Colorless to Yellow |
Density | 1.2000g/mL |
Flash Point | 110°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | CH3CONHCH2CH2Cl |
Refractive Index | 1.4770 to 1.48 |
Show More |
Safety and Handling
Product Identifier
- N-(2-Chloroethyl)acetamide
- Warning
- Specific target organ toxicity after repeated exposure Category 2
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'