missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(2-Chlorobenzyl)-2-(thiophen-2-yl)ethanamine Hydrochloride, Mikromol™
Description
N-(2-Chlorobenzyl)-2-(thiophen-2-yl)ethanamine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | N-(2-Chlorobenzyl)-2-(thiophen-2-yl)ethanamine Hydrochloride |
| CAS | 60612-23-9 |
| Analyte or Component Names | N-(2-Chlorobenzyl)-2-(thiophen-2-yl)ethanamine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H14 Cl N S . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Ticlopidine Hydrochloride Imp. I (EP), Ticlopidine Imp. I (EP), N-(2-Chlorobenzyl)-2-(thiophen-2-yl)ethanamine Hydrochloride, Ticlopidine Hydrochloride Imp. I (EP) as Hydrochloride, Ticlopidine Hydrochloride Impurity I as Hydrochloride, Ticlopidine Impurity I as Hydrochloride |
| InChI Formula | InChI=1S/C13H14ClNS.ClH/c14-13-6-2-1-4-11(13)10-15-8-7-12-5-3-9-16-12;/h1-6,9,15H,7-8,10H2;1H |
| SMILES | Cl.Clc1ccccc1CNCCc2cccs2 |
| IUPAC Name | N-[(2-chlorophenyl)methyl]-2-thiophen-2-ylethanamine;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?