missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Chlorobenzyl)-2-(2-thienyl)ethylamine Hydrochloride, TRC

Product Code. 30349463
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349463 100 mg 100mg
30354063 500 mg 500mg
30353848 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30349463 Supplier Toronto Research Chemicals Supplier No. TRCC375190100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60612-23-9
Molecular Formula C13 H14 Cl N S . Cl H
Molecular Weight (g/mol) 288.24
Synonym N-(2-Chlorobenzyl)-2-(thiophen-2-yl)ethanamine Hydrochloride, Ticlopidine Hydrochloride Imp. I (EP) as Hydrochloride
IUPAC Name N-[(2-chlorophenyl)methyl]-2-thiophen-2-ylethanamine;hydrochloride
SMILES Cl.Clc1ccccc1CNCCc2cccs2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H14ClNS.ClH/c14-13-6-2-1-4-11(13)10-15-8-7-12-5-3-9-16-12;/h1-6,9,15H,7-8,10H2;1H
Formula Weight 287.0302
Chemical Name or Material N-(2-Chlorobenzyl)-2-(thiophen-2-yl)ethanamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.