missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Chloro-6-methylphenyl)-2-[(6-chloro-2-methyl-4-pyrimidinyl)amino]-5-thiazolecarboxamide, TRC

Product Code. 30357849
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357849 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357849 Supplier Toronto Research Chemicals Supplier No. TRCC369290100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(2-Chloro-6-methylphenyl)-2-[(6-chloro-2-methylpyrimidin-4-yl)amino]thiazole-5-carboxamide
CAS 302964-08-5
Molecular Formula C16 H13 Cl2 N5 O S
Quantity 100 mg
Synonym 5-Thiazolecarboxamide, N-(2-chloro-6-methylphenyl)-2-[(6-chloro-2-methyl-4-pyrimidinyl)amino]-, N-(2-Chloro-6-methylphenyl)-2-[(6-chloro-2-methyl-4-pyrimidinyl)amino]-5-thiazolecarboxamide, 2-[(6-Chloro-2-methylpyrimidin-4-yl)amino]-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide
InChI Formula InChI=1S/C16H13Cl2N5OS/c1-8-4-3-5-10(17)14(8)23-15(24)11-7-19-16(25-11)22-13-6-12(18)20-9(2)21-13/h3-7H,1-2H3,(H,23,24)(H,19,20,21,22)
SMILES Cc1cccc(Cl)c1NC(=O)c2cnc(Nc3cc(Cl)nc(C)n3)s2
IUPAC Name N-(2-chloro-6-methylphenyl)-2-[(6-chloro-2-methylpyrimidin-4-yl)amino]-1,3-thiazole-5-carboxamide
Molecular Weight (g/mol) 394.28
Formula Weight 393.0218
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.