missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Chloro-4-pyrimidinyl)-2,3-dimethyl-2H-indazol-6-amine, TRC

Product Code. 30134037
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134037 10 g 10g
30004360 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134037 Supplier Toronto Research Chemicals Supplier No. TRCC38008010G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 444731-74-2
Molecular Formula C13 H12 Cl N5
Molecular Weight (g/mol) 273.7209
Synonym Pazopanib related, GW 790323X
IUPAC Name N-(2-chloropyrimidin-4-yl)-2,3-dimethylindazol-6-amine
SMILES Cc1c2ccc(Nc3ccnc(Cl)n3)cc2nn1C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C13H12ClN5/c1-8-10-4-3-9(7-11(10)18-19(8)2)16-12-5-6-15-13(14)17-12/h3-7H,1-2H3,(H,15,16,17)
Formula Weight 273.0781
Chemical Name or Material N-(2-chloropyrimidin-4-yl)-2,3-dimethylindazol-6-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.