missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Butynyl)phthalimide, 97%, Thermo Scientific™

Product Code. 15470408
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470408 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15470408 Supplier Thermo Scientific Alfa Aesar Supplier No. H53509.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 113439-83-1
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.21
MDL Number MFCD00160830
InChI Key COPWPECHFWIYSP-UHFFFAOYSA-N
Synonym n-2-butynyl phthalimide, 2-but-2-yn-1-yl isoindoline-1,3-dione, 2-but-2-yn-1-yl isoindole-1,3-dione, acmc-1bscb, 1-phthalimido-2-butyne, 4-phthalimido-2-butyne, 2-2-butynyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2-butyn-1-yl
PubChem CID 8063352
IUPAC Name 2-but-2-ynylisoindole-1,3-dione
SMILES CC#CCN1C(=O)C2=CC=CC=C2C1=O

Specifikationer

Melting Point 219°C to 223°C
Quantity 25 g
Formula Weight 199.21
Percent Purity 97%
Chemical Name or Material N-(2-Butynyl)phthalimide
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.