missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(2-Bromoethyl)phthalimide, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
N-(2-Bromoethyl)phthalimide is an intermediate used in organic synthesis. It can react with phenyl magnesium bromide to get 2-(2-bromo-ethyl)-3-hydroxy-3-phenyl-isoindolin-1-one.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-(2-Bromoethyl)phthalimide is an intermediate used in organic synthesis. It can react with phenyl magnesium bromide to get 2-(2-bromo-ethyl)-3-hydroxy-3-phenyl-isoindolin-1-one.
Solubility
Soluble in water (224 mg/L at 25°C).
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
N-(2-Bromoethyl)phthalimide is an intermediate used in organic synthesis. It can react with phenyl magnesium bromide to get 2-(2-bromo-ethyl)-3-hydroxy-3-phenyl-isoindolin-1-one.
Solubility
Soluble in water (224 mg/L at 25°C).
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
Specifications
Melting Point | 80°C to 84°C |
Quantity | 500 g |
Beilstein | 148736 |
Solubility Information | Soluble in water (224mg/L at 25°C). |
Formula Weight | 254.09 |
Percent Purity | ≥98% |
Chemical Name or Material | N- (2-Bromoethyl)phthalimide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'